1H-Indazole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Indazole-3-carboxylic acid

I0672-5G I0672-25G
TCI0672-5GEA 24 USD
TCI0672-5G TCI0672-25G
1H-Indazole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1H-Indazole-3-carboxylic acid

CAS Number: 4498-67-3
MDL Number: MFCD00211066
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 261


Other Product Names / Synonyms: 3-Carboxyindazole

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 261 °C
MDL Number: MFCD00211066
CAS Number: 4498-67-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fdcc12b4b80b06712fd2c08428a5d3128f9f10f&vid=4fdcc12b4b80b06712fd2c08428a5d3128f9f10f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR