(S)-(+)-2-(4-Isobutylphenyl)propionic Acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-2-(4-Isobutylphenyl)propanoic acid, (S)-(+)-4-Isobutyl-alpha-methylphenylacetic Acid, (S)-(+)-Ibuprofen

I0549-1G I0549-5G
TCI0549-001GEA 26.45 USD
TCI0549-001G TCI0549-005G
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid ≥98.0% (by GC, titration analysis)
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid

CAS Number: 51146-56-6
MDL Number: MFCD00069289
Molecular Formula: C13H18O2
Molecular Weight: 206.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 52
Flash Point (°C): 110
Specific rotation [a]20/D: 59 deg (C=2, EtOH)


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: (S)-(+)-Ibuprofen, (S)-(+)-4-Isobutyl-alpha-methylphenylacetic Acid

Formula: C₁₃H₁₈O₂
MW: 206.29 g/mol
Melting Pt: 52 °C
Flash Pt: 110 °C
MDL Number: MFCD00069289
CAS Number: 51146-56-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e0cc31fe19d00d4e0ac628e29bfda0efc7a4ef7&vid=8e0cc31fe19d00d4e0ac628e29bfda0efc7a4ef7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR