3-Isochromanone ≥99.0%
Supplier: TCI America
CAS Number: 4385-35-7
MDL Number: MFCD00043005
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 83
Other Product Names / Synonyms: 1,4-Dihydro-3H-2-benzopyran-3-one
Formula:
C₉H₈O₂ Boiling Pt: 130 °C Melting Pt: 83 °C |
MDL Number:
MFCD00043005 CAS Number: 4385-35-7 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...