2-Biphenylyl isocyanate ≥98.0%

Supplier: TCI America

Synonyms: 2-Biphenyl Isocyanate, Isocyanic acid 2-Biphenyl ester, 2-Biphenylylisocyanate

I0503-5G I0503-25G
TCI0503-005GEA 103.6 USD
TCI0503-005G TCI0503-25G
2-Biphenylyl isocyanate ≥98.0%
2-Biphenylyl isocyanate

CAS Number: 17337-13-2
MDL Number: MFCD00037087
Molecular Formula: C13H9NO
Molecular Weight: 195.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 109
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: Isocyanic Acid 2-Biphenyl Ester

Formula: C₁₃H₉NO
MW: 195.2167 g/mol
Boiling Pt: 130 °C
Density: 1.14 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00037087
CAS Number: 17337-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da52ae2d319304bafdc6ca6a0a086fab1201519c&vid=da52ae2d319304bafdc6ca6a0a086fab1201519c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR