4-(2-Iodoacetamido)-2,2,6,6-tetramethylpiperidinooxy ≥98.0% (by HPLC) free radical
Supplier: TCI America
Synonyms:
4-(2-Iodoacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl
, 4-(2-Iodoacetamido)-TEMPO
CAS Number: 25713-24-0
MDL Number: MFCD00010171
Molecular Formula: C11H20IN2O2
Molecular Weight: 339.20
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 127
Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-(2-Iodoacetamido)-TEMPO
Formula:
C₁₁H₂₀IN₂O₂ MW: 339.2 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD00010171 CAS Number: 25713-24-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...