Dimethyl-L-aspartate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
(S)-Aminosuccinic Acid Dimethyl Ester Hydrochloride, L-Asparaginic Acid Dimethyl Ester Hydrochloride, Dimethyl (S)-Aminosuccinate Hydrochloride, H-Asp(ome)-OMe HCl, Dimethyl L-Asparaginate Hydrochloride, H-Asp(OMe)-OMe.HCl, L-Aspartic Acid Dimethyl Ester Hydrochloride
CAS Number: 32213-95-9
MDL Number: MFCD00038878
Molecular Formula: C6H11NO4
Molecular Weight: 197.62
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 116
Specific rotation [a]20/D: 12 deg (C=1.4, H2O)
Product Categories: Bioscience, Amino Acids (C-Protected)
Formula:
C₆H₁₁NO₄ MW: 197.62 g/mol Melting Pt: 116 °C |
MDL Number:
MFCD00038878 CAS Number: 32213-95-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (N,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...