9-Acridinecarboxylic acid hydrate ≥97.0% (by HPLC)
Supplier: TCI America
A1505-1G
A1505-5G
TCA1505-001GEA
60.7
USD
TCA1505-001G
TCA1505-005G
9-Acridinecarboxylic acid hydrate ≥97.0% (by HPLC)
9-Acridinecarboxylic acid hydrate
CAS Number: 332927-03-4
MDL Number: MFCD00149578
Molecular Formula: C14H9NO2
Molecular Weight: 223.23
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Formula:
C₁₄H₁₁NO₃ |
MDL Number:
MFCD00149578 CAS Number: 332927-03-4 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...