Methyl (S)-(-)-2-isocyanato-4-methylvalerate ≥98.0%

Supplier: TCI America

Synonyms: (S)-(-)-2-Isocyanato-4-methylvaleric Acid Methyl Ester

I0467-5G I0467-25G
TCI0467-005GEA 464 USD
TCI0467-005G TCI0467-25G
Methyl (S)-(-)-2-isocyanato-4-methylvalerate ≥98.0%
Methyl (S)-(-)-2-isocyanato-4-methylvalerate

CAS Number: 39570-63-3
MDL Number: MFCD00191532
Molecular Formula: C8H13NO3
Molecular Weight: 171.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 74
Specific Gravity (20/20): 1.03


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-2-Isocyanato-4-methylvaleric Acid Methyl Ester

Formula: C₈H₁₃NO₃
MW: 171.20 g/mol
Boiling Pt: 74 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00191532
CAS Number: 39570-63-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=397415d007d54d835f437fdf3f67b0908daf66c5&vid=397415d007d54d835f437fdf3f67b0908daf66c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR