Isobutylene sulfide ≥98.0%

Supplier: TCI America

Synonyms: 2,2-Dimethylthiirane, 2-Methylpropylene sulphide, 2,2-Dimethylethylene Sulphide

I0461-5ML I0461-25ML
TCI0461-005MLEA 132.33 USD
TCI0461-005ML TCI0461-25ML
Isobutylene sulfide ≥98.0%
Isobutylene sulfide

CAS Number: 3772-13-2
MDL Number: MFCD00191662
Molecular Formula: C4H8S
Molecular Weight: 88.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: 2,2-Dimethylethylene Sulfide, 2,2-Dimethylthiirane, 2-Methylpropylene Sulfide

Formula: C₄H₈S
MW: 88.17 g/mol
Boiling Pt: 86 °C
Density: 0.90 (20 °C)
MDL Number: MFCD00191662
CAS Number: 3772-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62f147c1955eff78f2f878c02c194a303d1a9862&vid=62f147c1955eff78f2f878c02c194a303d1a9862&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR