4-Isobutylbenzaldehyde ≥95.0%

Supplier: TCI America

I0436-25ML I0436-500ML I0436-100ML
TCI0436-025MLEA 18.27 USD
TCI0436-025ML TCI0436-500ML TCI0436-100ML
4-Isobutylbenzaldehyde ≥95.0%
4-Isobutylbenzaldehyde

CAS Number: 40150-98-9
MDL Number: MFCD00075760
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.96

Formula: C₁₁H₁₄O
Density: 0.96 (20 °C)
MDL Number: MFCD00075760
CAS Number: 40150-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ade9acd4181e5e59797d833055f5b9ed1edbf262&vid=ade9acd4181e5e59797d833055f5b9ed1edbf262&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR