N-Isopropylacrylamide ≥98.0% stabilized

Supplier: TCI America

I0401-25G I0401-500G I0401-100G
TCI0401-025GEA 79.92 USD
TCI0401-025G TCI0401-500G TCI0401-100G
N-Isopropylacrylamide ≥98.0% stabilized
N-Isopropylacrylamide

(stabilized with MEHQ)
CAS Number: 2210-25-5
MDL Number: MFCD00041913
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 110
Melting point (°C): 66
Storage Temperature: 0-10°C

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 110 °C
Melting Pt: 66 °C
MDL Number: MFCD00041913
CAS Number: 2210-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7349923dba97ea85027d68ec7344f44c90fe85e5&vid=7349923dba97ea85027d68ec7344f44c90fe85e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR