N-Acetyl-2-(4-nitrophenyl)ethylamine ≥98.0%

Supplier: TCI America

Synonyms: N-[2-(4-Nitrophenyl)ethyl]acetamide, N-Acetyl-4-nitrophenethylamine

A1494-5G A1494-25G
TCA1494-5GEA 113 USD
TCA1494-5G TCA1494-025G
N-Acetyl-2-(4-nitrophenyl)ethylamine ≥98.0%
N-Acetyl-2-(4-nitrophenyl)ethylamine

CAS Number: 6270-07-1
MDL Number: MFCD00024805
Molecular Formula: C10H12N2O3
Molecular Weight: 208.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 140


Other Product Names / Synonyms: N-Acetyl-4-nitrophenethylamine, N-[2-(4-Nitrophenyl)ethyl]acetamide

Formula: C₁₀H₁₂N₂O₃
MW: 208.22 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00024805
CAS Number: 6270-07-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26ec9d4134705d9a338dcba66f856149f1bdfe61&vid=26ec9d4134705d9a338dcba66f856149f1bdfe61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR