2-Isopropenyltoluene ≥98.0% stabilized

Supplier: TCI America

Synonyms: 1-Methyl-2-(prop-1-en-2-yl)benzene

I0298-5ML I0298-1ML
TCI0298-005MLEA 761 USD
TCI0298-005ML TCI0298-1ML
2-Isopropenyltoluene ≥98.0% stabilized
2-Isopropenyltoluene

(stabilized with BHT)
CAS Number: 7399-49-7
MDL Number: MFCD00049092
Molecular Formula: C10H12
Molecular Weight: 132.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 43
Specific Gravity (20/20): 0.89

Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 173 °C
Density: 0.89 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD00049092
CAS Number: 7399-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93274d6ea7ed74f2b0b21d6ed965efee38186967&vid=93274d6ea7ed74f2b0b21d6ed965efee38186967&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR