Isobutyraldehyde diethyl acetal ≥96.0%

Supplier: TCI America

Synonyms: 1,1-Diethoxy-2-methylpropane, 1,1-Diethoxyisobutane

I0287-5ML I0287-25ML
TCI0287-005MLEA 76 USD
TCI0287-005ML TCI0287-025ML
Isobutyraldehyde diethyl acetal ≥96.0%
Isobutyraldehyde diethyl acetal

CAS Number: 1741-41-9
MDL Number: MFCD00059384
Molecular Formula: C8H18O2
Molecular Weight: 146.23
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: 1,1-Diethoxyisobutane

Formula: C₈H₁₈O₂
Boiling Pt: 138 °C
Density: 0.82 (20 °C)
MDL Number: MFCD00059384
CAS Number: 1741-41-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9582171b45a765f5b8849cee885139f222b4cf48&vid=9582171b45a765f5b8849cee885139f222b4cf48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR