Triallyl isocyanurate ≥96.0% (by GC) stabilized
Supplier: TCI America
|
Danger
|
Synonyms:
1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, Isocyanuric Acid Triallyl Ester, 1,3,5-Triallyl-1,3,5-triazinane-2,4,6-trione
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
(stabilized with BHT)
CAS Number: 1025-15-6
MDL Number: MFCD00006554
Molecular Formula: C12H15N3O3
Molecular Weight: 249.27
Purity/Analysis Method:
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Isocyanuric Acid Triallyl Ester
Formula:
C₁₂H₁₅N₃O₃ MW: 249.27 g/mol Boiling Pt: 140 °C (6 mmHg) (lit.) Flash Pt: 245 °C |
MDL Number:
MFCD00006554 CAS Number: 1025-15-6 |
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...