Isoproterenol sulfate dihydrate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Isoprenaline Sulfate

I0261-25G I0261-5G
TCI0261-025GEA 297 USD
TCI0261-025G TCI0261-5G
Isoproterenol sulfate dihydrate ≥98.0% (by HPLC, total nitrogen)
Isoproterenol sulphate dihydrate

CAS Number: 299-95-6
MDL Number: MFCD00013139
Molecular Formula: C11H17NO3
Molecular Weight: 520.59
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 83


Product Categories: Bioscience, Reagents for Life-style related Disease Models, Phenylethylamines
Other Product Names / Synonyms: Isoprenaline Sulfate

Formula: C₁₁H₁₇NO₃
MW: 520.59 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00013139
CAS Number: 299-95-6
EINECS: 206-085-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24e8aa76eab4114dc122058ad5699deb3e883feb&vid=ae14a4e1acf8dc76f83f30737df287e0bb16b644&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 6.5 to 7.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR