Isophthalic dihydrazide ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

I0257-25G I0257-500G I0257-100G
TCI0257-025GEA 40 USD
TCI0257-025G TCI0257-500G TCI0257-100G
Isophthalic dihydrazide ≥95.0% (by HPLC, titration analysis)
Isophthalic dihydrazide

CAS Number: 2760-98-7
MDL Number: MFCD00025117
Molecular Formula: C8H10N4O2
Molecular Weight: 194.19
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 227


Other Product Names / Synonyms: Isophthalohydrazide

Formula: C₈H₁₀N₄O₂
Melting Pt: 227 °C
MDL Number: MFCD00025117
CAS Number: 2760-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c412bfdf9b36fd245c1923768285ad73ec7b6923&vid=c412bfdf9b36fd245c1923768285ad73ec7b6923&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR