Isopropyl phosphate (mono and di ester mixture)
Supplier: TCI America
Synonyms:
Phosphoric Acid Isopropyl Ester
I0247-25ML
I0247-500ML
TCI0247-025MLEA
36
USD
TCI0247-025ML
TCI0247-500ML
Isopropyl phosphate (mono and di ester mixture)
Isopropyl phosphate (mono and di ester mixture)
MDL Number: MFCD00135965
Molecular Formula: C6H15O4P
Molecular Weight: 182.16
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 120
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: Phosphoric Acid Isopropyl Ester
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...