Isophorondiamine ≥99.0%
Supplier: TCI America
Synonyms:
1-Amino-3-aminomethyl-3,5,5-trimethylcyclohexane, Isophoronediamine
I0228-25ML
I0228-500ML
TCI0228-025MLEA
18.89
USD
TCI0228-025ML
TCI0228-500ML
Isophorondiamine ≥99.0%
Isophorondiamine
CAS Number: 2855-13-2
MDL Number: MFCD00019397
Molecular Formula: C10H22N2
Molecular Weight: 170.30
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 247
Melting point (°C): 10
Flash Point (°C): 117
Specific Gravity (20/20): 0.93
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1-Amino-3-aminomethyl-3,5,5-trimethylcyclohexane
Formula:
C₁₀H₂₂N₂ Boiling Pt: 247 °C Melting Pt: 10 °C Density: 0.93 (20 °C) Flash Pt: 117 °C |
MDL Number:
MFCD00019397 CAS Number: 2855-13-2 EINECS: 220-666-8 UN: 2289 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...