5-Isopropyl-m-xylene ≥85.0%
Supplier: TCI America
CAS Number: 4706-90-5
MDL Number: MFCD00026405
Molecular Formula: C11H16
Molecular Weight: 148.25
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: 3,5-Dimethylcumene, 3,5-Dimethylisopropylbenzene
Formula:
C₁₁H₁₆ Boiling Pt: 195 °C Density: 0.86 (20 °C) |
MDL Number:
MFCD00026405 CAS Number: 4706-90-5 |
|
Specification Test Results
Purity/Analysis Method | >85.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...