2-Ethylhexyl vinyl ether ≥97.0%

Supplier: TCI America

Synonyms: Isooctyl Vinyl Ether

I0148-10G I0148-25G
TCI0148-010GEA 132.9 USD
TCI0148-010G TCI0148-25G
2-Ethylhexyl vinyl ether ≥97.0%
2-Ethylhexyl vinyl ether

CAS Number: 103-44-6
MDL Number: MFCD00053800
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 57
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Isooctyl Vinyl Ether

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 177 °C
Density: 0.81 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00053800
CAS Number: 103-44-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5a2c71de2fdbe1fd3000df5b1f4a054aa4a0dd&vid=ba5a2c71de2fdbe1fd3000df5b1f4a054aa4a0dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR