Isobutyl vinyl ether ≥99.0% stabilized

Supplier: TCI America

I0099-25ML I0099-500ML
TCI0099-025MLEA 27 USD
TCI0099-025ML TCI0099-500ML
Isobutyl vinyl ether ≥99.0% stabilized
Isobutyl vinyl ether

(stabilized with KOH) CAS Number: 109-53-5 MDL Number: MFCD00008934 Molecular Formula: C6H12O Molecular Weight: 100.16 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 83 Flash Point (°C): -15 Specific Gravity (20/20): 0.77 Storage Temperature: 0-10°C

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 83 °C
Density: 0.77 (20 °C)
Flash Pt: –15 °C
MDL Number: MFCD00008934
CAS Number: 109-53-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=967ee416f2ff1144f1d54d606991d3b70088bb6f&vid=967ee416f2ff1144f1d54d606991d3b70088bb6f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR