3-Hydroxyxanthen-9-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

H1269-1G H1269-5G
TCH1269-1GEA 32.4 USD
TCH1269-1G TCH1269-5G
3-Hydroxyxanthen-9-one ≥98.0% (by HPLC, titration analysis)
3-Hydroxyxanthen-9-one

CAS Number: 3722-51-8
MDL Number: MFCD00604748
Molecular Formula: C13H8O3
Molecular Weight: 212.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 243

Formula: C₁₃H₈O₃
Melting Pt: 243 °C
MDL Number: MFCD00604748
CAS Number: 3722-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f892c568a86fd68f198f99b034b466128d4ee03&vid=2f892c568a86fd68f198f99b034b466128d4ee03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR