4-Hydroxy-1,2,2,6,6-pentamethylpiperidine ≥98.0%

Supplier: TCI America

Synonyms: 1,2,2,6,6-Pentamethylpiperidin-4-ol, 1,2,2,6,6-Pentamethyl-4-piperidinol

H1068-500G H1068-25G
TCH1068-500GEA 167 USD
TCH1068-500G TCH1068-25G
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine ≥98.0%
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine

CAS Number: 2403-89-6
Molecular Formula: C10H21NO
Molecular Weight: 171.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 76


Other Product Names / Synonyms: 1,2,2,6,6-Pentamethyl-4-piperidinol

Formula: C₁₀H₂₁NO
MW: 171.28 g/mol
Boiling Pt: 120 °C
Melting Pt: 76 °C
MDL Number: MFCD00191220
CAS Number: 2403-89-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e67aba661f59b95afd79e8608b77dfb9a6ea8089&vid=e67aba661f59b95afd79e8608b77dfb9a6ea8089&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR