Methyl-6-hydroxy-2-naphthoate ≥98.0%

Supplier: TCI America

Synonyms: 6-Hydroxy-2-naphthoic acid methyll ester, Methyl 6-hydroxy-2-naphthoate

H1053-5G H1053-25G
TCH1053-5GEA 59 USD
TCH1053-5G TCH1053-25G
Methyl-6-hydroxy-2-naphthoate ≥98.0%
Methyl 6-hydroxy-2-naphthoate

CAS Number: 17295-11-3
MDL Number: MFCD00100526
Molecular Formula: C12H10O3
Molecular Weight: 202.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 170


Other Product Names / Synonyms: 6-Hydroxy-2-naphthoic Acid Methyl Ester

Formula: C₁₂H₁₀O₃
Melting Pt: 170 °C
MDL Number: MFCD00100526
CAS Number: 17295-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8830ae000e1e126afa58b5c9f173a44426f96551&vid=8830ae000e1e126afa58b5c9f173a44426f96551&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR