N-Hydroxy-4-nitrophthalimide ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Nitrophthaloxime
H1036-5G
H1036-1G
TCH1036-5GEA
197.7
USD
TCH1036-5G
TCH1036-1G
N-Hydroxy-4-nitrophthalimide ≥98.0% (by HPLC)
N-Hydroxy-4-nitrophthalimide
CAS Number: 105969-98-0
MDL Number: MFCD01115771
Molecular Formula: C8H4N2O5
Molecular Weight: 208.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 169
Product Categories: Synthetic Organic Chemistry, Oxidation
Other Product Names / Synonyms: 4-Nitrophthaloxime
Formula:
C₈H₄N₂O₅ MW: 208.13 g/mol Melting Pt: 169 °C |
MDL Number:
MFCD01115771 CAS Number: 105969-98-0 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...