4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one ≥98.0%

Supplier: TCI America

Synonyms: Vanillylideneacetone, Dehydrozingerone, 4-(4-Hydroxy-3-methoxyphenyl)but-3-en-2-one

H0996-25G H0996-5G
TCH0996-25GEA 220.74 USD
TCH0996-25G TCH0996-5G
4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one ≥98.0%
4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one

CAS Number: 1080-12-2
MDL Number: MFCD00012210
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 129


Other Product Names / Synonyms: Dehydrozingerone, Vanillylideneacetone

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Melting Pt: 129 °C
MDL Number: MFCD00012210
CAS Number: 1080-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94dd731934acf2652dc330743a3e5c86f56eacdb&vid=94dd731934acf2652dc330743a3e5c86f56eacdb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR