2-(3-Hydroxypropyl)benzimidazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-(2-Benzimidazolyl)propanol, 3-(1H-Benzo[d]imidazol-2-yl)propan-1-ol, 2-Benzimidazolepropanol

H0988-25G H0988-5G
TCH0988-25GEA 237.5 USD
TCH0988-25G TCH0988-5G
2-(3-Hydroxypropyl)benzimidazole ≥98.0% (by titrimetric analysis)
2-(3-Hydroxypropyl)benzimidazole

CAS Number: 2403-66-9
MDL Number: MFCD00022688
Molecular Formula: C10H12N2O
Molecular Weight: 176.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 166


Other Product Names / Synonyms: 2-Benzimidazolepropanol, 3-(2-Benzimidazolyl)propanol

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00022688
CAS Number: 2403-66-9
EINECS: 219-290-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70b2b5b3c3fb3c0a0a1de14ee65fac68aeae966f&vid=01822cf5f0dc3d42231efaf99be5af0aa70f3e32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR