2-Hexyl-4-pentynoic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(2-Propynyl)octanoic Acid, 2-Propargyloctanoic Acid

H0964-25G H0964-5G
TCH0964-025GEA 1003 USD
TCH0964-025G TCH0964-005G
2-Hexyl-4-pentynoic acid ≥95.0% (by titrimetric analysis)
2-Hexyl-4-pentynoic acid

CAS Number: 96017-59-3
MDL Number: MFCD00946863
Molecular Formula: C11H18O2
Molecular Weight: 182.26
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 0.94


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 2-Propargyloctanoic Acid, 2-(2-Propynyl)octanoic Acid

Formula: C₁₁H₁₈O₂
MW: 182.26 g/mol
Density: 0.94 (20 °C)
MDL Number: MFCD00946863
CAS Number: 96017-59-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4921f2a2791447fdec3f813186fba03b6479d3a0&vid=4921f2a2791447fdec3f813186fba03b6479d3a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR