(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone ≥94.0%
Supplier: TCI America
Synonyms:
(5S)-(+)-5-(Hydroxymethyl)dihydrofuran-2(3H)-one, (S)-5-(Hydroxymethyl)dihydrofuran-2(3H)-one, (S)-(+)-4,5-Dihydro-5-hydroxymethyl-2(3H)-furanone, (S)-(+)-5-Hydroxymethyltetrahydro-2-furanone
CAS Number: 32780-06-6
MDL Number: MFCD00066224
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific rotation [a]20/D: 57 deg (C=3, CHCl3)
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-4,5-Dihydro-5-hydroxymethyl-2(3H)-furanone, (S)-(+)-5-Hydroxymethyltetrahydro-2-furanone
Formula:
C₅H₈O₃ MW: 116.12 g/mol Flash Pt: 110 °C |
MDL Number:
MFCD00066224 CAS Number: 32780-06-6 |
Specification Test Results
Purity/Analysis Method | >94.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...