3-Hydroxyaspartic acid ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: 2-Amino-3-hydroxysuccinic Acid

H0947-5G H0947-1G
TCH0947-005GEA 293 USD
TCH0947-005G TCH0947-1G
3-Hydroxyaspartic acid ≥98.0% (by total nitrogen and titration analysis)
3-Hydroxyaspartic acid

CAS Number: 71653-06-0
MDL Number: MFCD00128126
Molecular Formula: C4H7NO5
Molecular Weight: 149.10
Purity/Analysis Method: >98.0% (T,N)
Form: Crystal


Other Product Names / Synonyms: 2-Amino-3-hydroxysuccinic Acid

Formula: C₄H₇NO₅
MW: 149.10 g/mol
MDL Number: MFCD00128126
CAS Number: 71653-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed136794109ceb0f2308b1fff094fa6d8a770a2c&vid=ed136794109ceb0f2308b1fff094fa6d8a770a2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR