3-Hydroxy-1,2-dimethyl-4(1H)-pyridone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0944-25G H0944-5G
TCH0944-025GEA 231 USD
TCH0944-025G TCH0944-005G
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone ≥98.0% (by titrimetric analysis)
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone

CAS Number: 30652-11-0
MDL Number: MFCD00134497
Molecular Formula: C7H9NO2
Molecular Weight: 139.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 260

Formula: C₇H₉NO₂
Melting Pt: 260 °C
MDL Number: MFCD00134497
CAS Number: 30652-11-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=927ede61ffdc5b5efcf8f995260f929140b4ba92&vid=927ede61ffdc5b5efcf8f995260f929140b4ba92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR