Ethyl-2-(hydroxymethyl)acrylate ≥95.0% stabilized

Supplier: TCI America

Synonyms: 2-(Hydroxymethyl)acrylic acid ethyl ester, Ethyl 2-(hydroxymethyl)acrylate

H0916-5G H0916-25G
TCH0916-005GEA 99 USD
TCH0916-005G TCH0916-25G
Ethyl-2-(hydroxymethyl)acrylate ≥95.0% stabilized
Ethyl-2-(hydroxymethyl)acrylate

(stabilized with HQ)
CAS Number: 10029-04-6
MDL Number: MFCD01673859
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Specific Gravity (20/20): 1.07
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-(Hydroxymethyl)acrylic Acid Ethyl Ester

Formula: C₆H₁₀O₃
MW: 130.1418 g/mol
Boiling Pt: 80 °C
Density: 1.07 (20 °C)
MDL Number: MFCD01673859
CAS Number: 10029-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91ff1acb07da3ebc13acd080f2200bc9323a18f5&vid=91ff1acb07da3ebc13acd080f2200bc9323a18f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR