Haloperidol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4'-fluorobutyrophenone

H0912-25G H0912-5G
TCH0912-025GEA 246 USD
TCH0912-025G TCH0912-5G
Haloperidol ≥98.0% (by HPLC, titration analysis)
Haloperidol

CAS Number: 52-86-8
MDL Number: MFCD00051423
Molecular Formula: C21H23ClFNO2
Molecular Weight: 375.87
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 152


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Inhibitors (for Pharmacological Research), Neurotransmitter Agent Ingredients, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4'-fluorobutyrophenone

Formula: C₂₁H₂₃ClFNO₂
MW: 375.87 g/mol
Melting Pt: 152 °C
MDL Number: MFCD00051423
CAS Number: 52-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcc1e978346c32c68ac3555c59b3fa05f11bdc6a&vid=bcc1e978346c32c68ac3555c59b3fa05f11bdc6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR