2-Hydroxy-3-butynoic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
H0905-1G
H0905-100MG
TCH0905-1GEA
1342.2
USD
TCH0905-1G
TCH0905-100MG
2-Hydroxy-3-butynoic acid ≥97.0% (by titrimetric analysis)
2-Hydroxy-3-butynoic acid
CAS Number: 38628-65-8
MDL Number: MFCD00671546
Molecular Formula: C4H4O3
Molecular Weight: 100.07
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Storage Temperature: <0°C
Formula:
C₄H₄O₃ MW: 100.07 g/mol |
MDL Number:
MFCD00671546 CAS Number: 38628-65-8 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...