3-(tert-Butoxycarbonylamino)-1-propanol ≥96.0%

Supplier: TCI America

Synonyms: 3-(Boc-amino)-1-propanol, N-(3-Hydroxypropyl)carbamic Acid tert-Butyl Ester, Boc-β-Ala-ol, 3-(BOC-amino)-1-propanol, tert-Butyl N-(3-Hydroxypropyl)carbamate

H0900-25ML H0900-5ML
TCH0900-025MLEA 117.99 USD
TCH0900-025ML TCH0900-005ML
3-(tert-Butoxycarbonylamino)-1-propanol ≥96.0%
3-(tert-Butoxycarbonylamino)-1-propanol

CAS Number: 58885-58-8
MDL Number: MFCD00191883
Molecular Formula: C8H17NO3
Molecular Weight: 175.23
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 109
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 3-(Boc-amino)-1-propanol, tert-Butyl N-(3-Hydroxypropyl)carbamate, N-(3-Hydroxypropyl)carbamic Acid tert-Butyl Ester

Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
Density: 1.03 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00191883
CAS Number: 58885-58-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=360c981af42765a58a7acb48453ceee35fae1f52&vid=360c981af42765a58a7acb48453ceee35fae1f52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR