4-Methyl-1-acetoxycalixarene [mixture of [6] and [8]] ≥95.0% (by HPLC) (contains 5-10% Acetone)
Supplier: TCI America
Synonyms:
Hexamethyl(hexaacetoxy)calixarene, 1-Acetoxy-4-methylcalixarene
H0889-1G
H0889-100MG
TCH0889-001GEA
395.4
USD
TCH0889-001G
TCH0889-100MG
4-Methyl-1-acetoxycalixarene [mixture of [6] and [8]] ≥95.0% (by HPLC) (contains 5-10% Acetone)
4-Methyl-1-acetoxycalixarene [mixture of [6] and [8]]
MDL Number: MFCD00671543
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Other Product Names / Synonyms: 1-Acetoxy-4-methylcalixarene, Hexamethyl(hexaacetoxy)calixarene
Specification Test Results
Purity/Analysis Method | >95.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...