6-Hydroxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-Hydroxy-1-naphthalenecarboxylic acid

H0886-5G H0886-25G
TCH0886-5GEA 291 USD
TCH0886-5G TCH0886-025G
6-Hydroxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)
6-Hydroxy-1-naphthoic acid

CAS Number: 2437-17-4
MDL Number: MFCD00209983
Molecular Formula: C11H8O3
Molecular Weight: 188.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 210


Other Product Names / Synonyms: 6-Hydroxy-1-naphthalenecarboxylic Acid

Formula: C₁₁H₈O₃
MW: 188.1800 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00209983
CAS Number: 2437-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adcc40b08be63dcbda0c6049ead27a1a5c3ccb45&vid=adcc40b08be63dcbda0c6049ead27a1a5c3ccb45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR