Methyl-5-hexenoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 5-hexenoate, 5-Hexenoic Acid Methyl Ester

H0872-5G H0872-25G
TCH0872-005GEA 203.9 USD
TCH0872-005G TCH0872-025G
Methyl-5-hexenoate ≥98.0%
Methyl-5-hexenoate

CAS Number: 2396-80-7
MDL Number: MFCD00671538
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 46
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: 5-Hexenoic Acid Methyl Ester

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 157 °C
Density: 0.91 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00671538
CAS Number: 2396-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=157ff33756a8a28ee6b7027503370c708343645a&vid=157ff33756a8a28ee6b7027503370c708343645a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR