Acetoxime O-(2,4,6-Trimethylphenylsulfonate) ≥98.0% (by HPLC)
Supplier: TCI America
CAS Number: 81549-07-7
MDL Number: MFCD01321127
Molecular Formula: C12H17NO3S
Molecular Weight: 255.33
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 96
Product Categories: Synthetic Organic Chemistry, Amination
Other Product Names / Synonyms: Acetone O-(2,4,6-Trimethylphenylsulfonyl)oxime, O-(2,4,6-Trimethylphenylsulfonyl)acetoxime
Formula:
C₁₂H₁₇NO₃S Melting Pt: 96 °C |
MDL Number:
MFCD01321127 CAS Number: 81549-07-7 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...