ɑ,ɑ,ɑ',ɑ',4,5-Hexabromo-o-xylene ≥98.0%

Supplier: TCI America

H0831-5G H0831-1G
TCH0831-005GEA 328.1 USD
TCH0831-005G TCH0831-1G
ɑ,ɑ,ɑ',ɑ',4,5-Hexabromo-o-xylene ≥98.0%
alpha,alpha,alpha',alpha',4,5-Hexabromo-o-xylene

CAS Number: 13209-20-6
MDL Number: MFCD00191445
Molecular Formula: C8H4Br6
Molecular Weight: 579.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 135

Formula: C₈H₄Br₆
MW: 579.54 g/mol
Melting Pt: 135 °C
MDL Number: MFCD00191445
CAS Number: 13209-20-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3562ba39cc9ee5bc29f2550f452976fc8e04b6e9&vid=3562ba39cc9ee5bc29f2550f452976fc8e04b6e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR