4-Hydroxy-4'-methoxybiphenyl ≥95.0%
Supplier: TCI America
Synonyms:
4-Hydroxy-4'-methoxydiphenyl, 4'-Methoxy-[1,1'-biphenyl]-4-ol, 4'-Methoxybiphenyl-4-ol
H0819-1G
H0819-5G
TCH0819-001GEA
73
USD
TCH0819-001G
TCH0819-005G
4-Hydroxy-4'-methoxybiphenyl ≥95.0%
4-Hydroxy-4'-methoxybiphenyl
CAS Number: 16881-71-3
MDL Number: MFCD00191514
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 181
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Hydroxy-4'-methoxydiphenyl, 4'-Methoxybiphenyl-4-ol
Formula:
C₁₃H₁₂O₂ MW: 200.24 g/mol Melting Pt: 181 °C |
MDL Number:
MFCD00191514 CAS Number: 16881-71-3 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...