Acetaldehyde ammonia trimer ≥95.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 58052-80-5
MDL Number: MFCD00149559
Molecular Formula: C6H15N3
Molecular Weight: 129.21
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 95
Flash Point (°C): 55
Other Product Names / Synonyms: Aldehyde Ammonia Trimer, Hexahydro-2,4,6-trimethyl-1,3,5-triazine Trihydrate, 2,4,6-Trimethylhexahydro-1,3,5-triazine Trihydrate
Formula:
C₆H₂₁N₃O₃ Melting Pt: 95 °C Flash Pt: 55 °C |
MDL Number:
MFCD00149559 CAS Number: 58052-80-5 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...