Hexadecyltrimethylammonium perchlorate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

H0735-1G H0735-5G
TCH0735-001GEA 98.3 USD
TCH0735-001G TCH0735-5G
Hexadecyltrimethylammonium perchlorate ≥98.0% (by total nitrogen basis)
Hexadecyltrimethylammonium perchlorate

CAS Number: 6941-37-3
MDL Number: MFCD00144895
Molecular Formula: C19H42ClNO4
Molecular Weight: 384.00
Purity/Analysis Method: >98.0% (N)
Form: Crystal


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Cetyltrimethylammonium Perchlorate

Formula: C₁₉H₄₂ClNO₄
MDL Number: MFCD00144895
CAS Number: 6941-37-3

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR