Hexadecyltrimethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

H0734-5G H0734-1G
TCH0734-5GEA 335.8 USD
TCH0734-5G TCH0734-001G
Hexadecyltrimethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Hexadecyltrimethylammonium tetrafluoroborate

CAS Number: 73257-08-6
MDL Number: MFCD00144896
Molecular Formula: C19H42BF4N
Molecular Weight: 371.36
Purity/Analysis Method: >98.0% (N)
Form: Crystal


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Cetyltrimethylammonium Tetrafluoroborate

Formula: C₁₉H₄₂BF₄N
MDL Number: MFCD00144896
CAS Number: 73257-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=baa1aed9c1631f020dfdf258d0cb615455c1d4a5&vid=baa1aed9c1631f020dfdf258d0cb615455c1d4a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR