9-Trifluoroacetylanthracene ≥98.0%

Supplier: TCI America

Synonyms: 9-Anthryl Trifluoromethyl Ketone

A1422-5G A1422-1G
TCA1422-005GEA 216 USD
TCA1422-005G TCA1422-001G
9-Trifluoroacetylanthracene ≥98.0%
9-Trifluoroacetylanthracene

CAS Number: 53531-31-0
MDL Number: MFCD00001258
Molecular Formula: C16H9F3O
Molecular Weight: 274.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 9-Anthryl Trifluoromethyl Ketone

Formula: C₁₆H₉F₃O
MW: 274.24 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00001258
CAS Number: 53531-31-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8362b73ccf0b18e007682808728c977c820e198&vid=d8362b73ccf0b18e007682808728c977c820e198&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR