Methyl-4-(2-hydroxyethoxy)salicylate ≥98.0%

Supplier: TCI America

Synonyms: 4-(2-Hydroxyethoxy)salicylic Acid Methyl Ester2-Hydroxy-4-(2-hydroxyethoxy)benzoic Acid Methyl EsterMethyl 2-Hydroxy-4-(2-hydroxyethoxy)benzate

H0727-5G H0727-1G
TCH0727-1GEA 84 USD
TCH0727-1G TCH0727-005G
Methyl-4-(2-hydroxyethoxy)salicylate ≥98.0%
Methyl-4-(2-hydroxyethoxy)salicylate

MDL Number: MFCD00143216
Molecular Formula: C10H12O5
Molecular Weight: 212.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 85


Other Product Names / Synonyms: 4-(2-Hydroxyethoxy)salicylic Acid Methyl Ester, Methyl 2-Hydroxy-4-(2-hydroxyethoxy)benzate, 2-Hydroxy-4-(2-hydroxyethoxy)benzoic Acid Methyl Ester

https://pr.vwr.com/stibo/web/std.lang.all/07/66/28970766.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR