6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid ≥98.0%
Supplier: TCI America
Synonyms:
Trolox, 6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid
H0726-5G
H0726-1G
TCH0726-5GEA
307.28
USD
TCH0726-5G
TCH0726-001G
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid ≥98.0%
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid
CAS Number: 53188-07-1
MDL Number: MFCD00006846
Molecular Formula: C14H18O4
Molecular Weight: 250.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Product Categories: Bioscience, Antioxidants, ROS/RNS Detection Reagents, Vitamin Derivatives, Redox Research Reagents
Other Product Names / Synonyms: Trolox
Formula:
C₁₄H₁₈O₄ MW: 250.29 g/mol |
MDL Number:
MFCD00006846 CAS Number: 53188-07-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...