2-Hydroxy-5-methylisophthalaldehyde ≥99.0%

Supplier: TCI America

Synonyms: 2-Hydroxy-5-methylbenzene-1,3-dicarbaldehyde

H0683-1G H0683-5G
TCH0683-001GEA 252 USD
TCH0683-001G TCH0683-5G
2-Hydroxy-5-methylisophthalaldehyde ≥99.0%
2-Hydroxy-5-methylisophthalaldehyde

CAS Number: 7310-95-4
MDL Number: MFCD00016596
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 132

Formula: C₉H₈O₃
MW: 164.1600 g/mol
Melting Pt: 132 °C
MDL Number: MFCD00016596
CAS Number: 7310-95-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c94cc9c8ee13f7275cdcc2d64d07fd261bbc5eb1&vid=c94cc9c8ee13f7275cdcc2d64d07fd261bbc5eb1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR