2-Hydroxy-2-methylbutyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Hydroxy-2-methylbutanoic acid

H0636-5G H0636-25G
TCH0636-005GEA 58 USD
TCH0636-005G TCH0636-25G
2-Hydroxy-2-methylbutyric acid ≥98.0% (by titrimetric analysis)
2-Hydroxy-2-methylbutyric acid

CAS Number: 3739-30-8
MDL Number: MFCD00004476
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 109
Melting point (°C): 72

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 109 °C
Melting Pt: 72 °C
MDL Number: MFCD00004476
CAS Number: 3739-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7127121b76dfabcbdd9dfabbc4014a59394ae715&vid=7127121b76dfabcbdd9dfabbc4014a59394ae715&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR